Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP28 Rabbit Polyclonal Antibody

Supplier: Proteintech
17707-1-AP-150UL
10096-860EA 453 USD InStock
10096-860
Anti-USP28 Rabbit Polyclonal Antibody
Antibodies
USP28(Ubiquitin carboxyl-terminal hydrolase 28) is also named as KIAA1515. USP28 is a 53BP1-interacting protein and a putative deubiquitinating enzyme, and belongs to a family of enzymes, called deubiquitinating enzymes (DUBs), which remove ubiquitin from the ubiquitin conjugates. It can be phosphorylated upon DNA damage at Ser-67 and Ser-714, by ATM or ATR. This protein has 2 isoforms produced by alternative splicing with the molecular mass of 122 kDa and 119 kDa.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: USP28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty