Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZNF238 Rabbit Polyclonal Antibody

Supplier: Proteintech
12714-1-AP-150UL
10097-320EA 453 USD InStock
10097-320
Anti-ZNF238 Rabbit Polyclonal Antibody
Antibodies
ZNF238 is a member of the BTB/POZ-ZF protein family, which involve in development and cancer formation, for example BCL-6, PLZF, and HIC-1. It's a transcriptional repressor involve in myogenesis and brain development. By directly repressing the expression of two skeletal myogenesis inhibitors, ID2 and ID3, ZNF238 plays a key role in myogenesis. It can control cell division of progenitor cells and regulating the survival of postmitotic cortical neurons. Besides, ZNF238 involves in the organization of nuclear chromosomes, for its specific binding to the consensus DNA sequence that contains the E box core, and recruiting chromatin remodeling multiportein complex. ZNF238 proteins has apparent molecular masses of 60 and 48 kD. Specific binding is found for a 60-kDa band which corresponds to the full length of RP58 protein. In addition, a 48-kDa band, thought to be the truncated form 2 is detected .

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IP: Mouse cerebellum Tissue, 1:500-1:5000; IHC: Mouse Brain Tissue, 1:20-1:200

Type: Primary
Antigen: ZNF238
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty