Would you like to log in again?
Human ABCD1 ELISA Kit, Proteintech
Supplier: Proteintech
KE00030-96T
10097-434EA 337 USD InStock
10097-434
Human ABCD1 ELISA Kit, Proteintech
Assays ELISAs
Human ABCD1 ELISA kit is a solid phase sandwich enzyme linked-immuno-sorbent assay (sandwich ELISA) used to detect and quantify protein levels of endogenous ABCD1.
The assay recognizes human ABCD1. A polyclonal antibody specific for ABCD1 has been pre-coated onto the microwells. The ABCD1 protein in samples is captured by the coated antibody after incubation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty