Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL6 Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
XP-5195BT
10098-142EA 593.41 USD InStock
10098-142
Anti-IL6 Rabbit Polyclonal Antibody (Biotin)
Antibodies
IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. It acts upon a variety of cells including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. In addition, IL-6 appears to interact with IL2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 is a multifunctional 24 kDa protein originally discovered in the medium of RNA stimulated fibroblastoid cells. It is upregulated by IL1, TNF, PDGF, IFN beta, TNF alpha, NGF, IL17 and downregulated by glucocorticoids IL4, TGF beta. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be as important as IL1 and TNF alpha in regulating the acute phase response. IL6 has also been implicated in regulating adipose mass.

ELISA: To detect rat IL-6 by direct ELISA (using 100 µL/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 µg/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant rat IL-6. Sandwich: To detect Rat IL-6 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Rat IL-6 (XP-5195) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant Rat IL-6. Western Blot: To detect rat IL-6 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant rat IL-6 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

Anti-rat IL-6 specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Il6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty