Would you like to log in again?
ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Supplier: Eppendorf
ep Dualfilter T.I.P.S.® SealMax®
0030078730 0030078721 0030078748 0030078748 0030078713 0030078713 0030078691 0030078705 0030078705 0030078730 0030078691
10098-568CS 244.16 USD InStock
CA10067-766 10098-568 10098-578 CA10067-764 CA10067-774 CA10067-772 10098-572 10098-570 10098-576 10098-574 CA10067-768
ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Pipette Tips
Eppendorf ep Dualfilter T.I.P.S.® SealMax® Racks provide advanced self-sealing pipette tips in 'PCR clean and Sterile' grade, featuring unique two-phase filters for unparalleled protection against contamination in sensitive lab applications.
  • Reliable protection from liquids, aerosols, and biomolecules
  • No PCR inhibiting additives
  • Protection of pipette from aggressive liquids when over-aspiration occurs
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 20% lower polypropylene consumption per rack
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
Superior protection for both sample and pipettes. The ep Dualfilter T.I.P.S.® SealMax® self-sealing filter pipette tips offer almost 100 % protection from contaminating aerosols and biomolecules with a two-phase design that works together to ensure optimal pipetting. Over-pipetted samples are stopped in their tracks with the instant closure of the Dualfilter. The white layer retains drops and splashes while the violet layer forms a barrier.
ep Dualfilter T.I.P.S.® SealMax® are ideally suited for methods where high sensitivity and reproducibility are essential. Both ep Dualfilter T.I.P.S.® and ep Dualfilter T.I.P.S.® SealMax® are especially recommended for applications such as PCR and microbiology, where preserving sample integrity and preventing cross contamination are critical.
The ep Dualfilter T.I.P.S.® SealMax® self-sealing filter pipette tips are manufactured to the highest quality under cleanroom conditions, without the use of interfering slip agents, biocides, and plasticizers. Made from the purest, non-recycled materials, self-sealing filter pipette tips are ideal for molecular and microbiological applications that require the greatest sensitivity and reproducibility, without the fear of sample degradation and cross-contamination.
Certifications: Lot-specific certficates for the purity grade PCR clean/Sterile can be downloaded from Eppendorf website. Certificate of filter efficiency of class ISO 25 E in accordance with DIN EN ISO 29463-5 available.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty