Would you like to log in again?
GelRed Prestain Loading Buffers with Tracking Dye (6X), Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
41010
10098-684EA 125.82 USD InStock
10098-684
GelRed Prestain Loading Buffers with Tracking Dye (6X), Biotium
Buffers Electrophoresis Buffers Loading Buffers
GelRed is a sensitive, stable and environmentally safe fluorescent nucleic acid dyes are DNA gel loading buffers designed to replace the highly toxic ethidium bromide for one-step fluorescent staining and gel loading of DNA.
GelRed® and EtBr have virtually the same spectra, so you can directly replace EtBr with GelRed® without changing your existing imaging system. In addition, GelRed® is far more sensitive than EtBr, which cannot be used in DNA loading buffer to prestain DNA. GelRed® is compatible with downstream applications such as sequencing and cloning. GelRed® is efficiently removed from DNA by commercial gel extraction kits or by phenol/chloroform extraction and ethanol precipitation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty