Would you like to log in again?
Two-Piece Jacketed Pressure Reactor without Drain Valve, Ace Glass Incorporated
Supplier: Ace Glass
6427-417 6427-423 6427-316 6427-26 6427-323 6427-223 6427-272 6427-428 6427-18 6427-228 6427-270 6427-13 6427-207 6427-217 6427-209 6427-412 6427-313 6427-326 6427-268 6427-409 6427-16 6427-09 6427-264 6427-318 6427-266 6427-23 6427-274 6427-407 6427-212 6427-309
10767-776EA 17288.76 USD InStock
10767-776 10767-774 10098-838 10767-778 10098-844 89054-316 89054-338 89054-318 10098-842 10098-846 89054-330 89054-332 89054-310 10098-840 89054-312 89054-334 89054-314 89054-336 89054-326 10767-274 89054-328 10767-772 10098-836 10767-770 89054-340 89054-342 89054-320 89054-344 89054-322 89054-324
Two-Piece Jacketed Pressure Reactor without Drain Valve, Ace Glass Incorporated
Laboratory Reactor Systems
Complete, two-piece, jacketed flask pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis, and catalysis. Reactor capacities range from 500mL to 5000mL, and use Ace-Threds to achieve a leak-tight system. Two-piece (Duran® flanged flask and head) design allows the use of large agitators and facilitates product removal and clean-out. Systems can be selected with either a shaft and agitator style mixing, or stir bar mixing with the use of a super magnet suspended below the reactor body. Reactor bodies are available with 28/15 ball joint or Ace-Thred inlet and outlets. Does not include stand.
Rated 36 psi at 100 °C. Maximum jacket pressure is 8 psig.
Reactor capacities range from 500 mL to 5000 mL, and use Ace-Threds to achieve a leak-tight system. Two-piece (Duran® flanged flask and head) design allows the use of large agitators and facilitates product removal and clean-out. Systems can be selected with either a shaft and agitator style mixing, or stir bar mixing with the use of a super magnet suspended below the reactor body. Reactor bodies are available with 28/15 ball joint or Ace-Thred inlet and outlets. Does not include stand.
Ordering information: System is supplied complete with (2) #15 PTFE plugs, condenser, flask, head, manifold, thermowell, clamp, funnel, (2) #15 PTFE bushings, bearing with Ace-Threds, stir shaft, flexible shaft, PTFE stirrer blade, Krytox high vacuum grease, swivel coupling, polypropylene tubing, motor controller and selected mixing type, mounting rod, and PTFE sealing tape. Stir bar style systems come with raised ring octaganol stirrer magnet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty