Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Scale Up Series™ Jacketed Cylindrical Reactor Flasks, Ace Glass
Supplier: Ace Glass
6441-12 6441-10 6441-14 6441-02 6441-06 6441-04 6441-18 6441-16 6441-08
10098-910EA 7334.59 USD InStock
10098-910 10098-898 10098-914 10098-902 10098-912 10098-900 10098-906 10098-904 10098-908
Scale Up Series™ Jacketed Cylindrical Reactor Flasks, Ace Glass
Reaction Vessels Reaction Vessels, Flask Shape
Scale-Up Series™ Jacketed Cylindrical Flasks are equipped with an Easy Drain glass valve and KF/NW inlet and outlet
The flask is graduated on both sides, which allows for either left- or right-handed flask orientation. The glass plug Easy Drain valve is designed to prevent accidental valve removal when opening, and incorporates a compression tube angled side arm for maximum unobstructed drainage. Flasks are available in sizes ranging from 100mL to 6000mL. Operating temperature limits are –60 to 200°C (–76 to 392°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty