Would you like to log in again?
Rod Mounted Swing Latch Clamps, Ace Glass
Supplier: Ace Glass
6442-04 6442-06 6442-02
10098-920EA 905.83 USD InStock
10098-920 10098-918 10098-916
Rod Mounted Swing Latch Clamps, Ace Glass
Clamps General Purpose Clamps
Swing latch clamp secures neck of Protean Fusion reaction vessel to the stand.
May also be used to support any round object with an equivalent maximum outside diameter. Stainless steel rod support is 1/2" O.D×8" long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty