Would you like to log in again?
Jacketed Reaction Flasks, Ace Glass
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6519-302 6519-03 6519-04 6519-01 6519-304 6519-02 6519-301 6519-303
10098-968EA 1279.82 USD InStock
10098-968 89215-020 10098-966 89215-024 10098-972 89215-016 89215-006 10098-970
Jacketed Reaction Flasks, Ace Glass
Reaction Vessels Reaction Vessels, Flask Shape
This is a cylindrical, heavy-wall reaction flask, except flat flange has an O-ring groove for use with CAPFE® (PTFE encapsulated silicone rubber) O-ring instead of a gasket
Without constriction at top to facilitate introduction/removal of material and allow for ease of cleaning. Flat bottom rounded into side wall. Flask is supplied with one CAPFE O-ring. Comes with 28/15 O‑ring ball joint for easy connect/disconnect from hoses, or #11 Ace-Thred. Made from borosilicate glass.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty