Would you like to log in again?
Powder Funnels, Ace Glass
Supplier: Ace Glass
7236-18 7236-11 9488-10 7236-08 7236-10 7236-129 7236-20 7236-124 7236-145 7236-12 7236-06 7236-14 7236-16
89058-616EA 99.33 USD InStock
89058-616 10099-026 89058-618 10099-028 89074-940 10099-030 10099-032 89083-988 89058-620 89058-622 89058-624 89058-626 89058-614
Powder Funnels, Ace Glass
Funnels Powder Funnels
Useful in pouring powders or liquids into ground joint containers
These powder funnels are made of glass.
Available with standard taper or spherical inner joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty