Would you like to log in again?
Septa for GL Caps, Silicone and Silicone/PTFE, Ace Glass Incorporated
Supplier: Ace Glass
7625-04 7625-02 7625-05 7625-09 7625-06 7625-07 7625-10 7625-01 7625-08 7625-03
10099-412PK 89.91 USD InStock
10099-412 10099-106 10099-108 10099-404 10099-406 10099-408 10099-110 10099-410 10099-036 10099-112
Septa for GL Caps, Silicone and Silicone/PTFE, Ace Glass Incorporated
Septa
These septa are designed to fit the DURAN® open-top cap product family.
Available in both silicone and PTFE-faced silicone for good chemical resistance. Caps can withstand 130 °C (266 °F) vapor or 200 °C (392 °F) dry heat.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty