Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Labjaws Lab-Frame Aluminum Rods, Ace Glass Incorporated
Supplier: Ace Glass
11166-29 11166-27 11166-23 11166-22 11166-39 11166-33 11166-21 11166-25 11166-26 11166-37 11166-31
89084-724EA 24.94 USD InStock
89084-724 89084-734 89084-722 89084-732 10099-138 89084-730 89084-740 89084-728 89084-738 89084-726 89084-736
Labjaws Lab-Frame Aluminum Rods, Ace Glass Incorporated
Support Rods and Frames
Labjaws aluminium rods are ground to a 13 mm (1/2") diameter and are available in a variety of lengths to fit any custom lab frame setup.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty