Would you like to log in again?
PTFE Sleeves, 0.05 mm, Ace Glass Incorporated
Supplier: Ace Glass
7643-14 7643-29 7643-114 7643-125 7643-110 7643-20 7643-104 7643-12 7643-08 7643-112 7643-09 7643-106 7643-06 7643-120 7643-04 7643-16 7643-107 7643-116 7643-129 7643-109 7643-02 7643-102 7643-25 7643-118 7643-03 7643-18 7643-10 7643-103 7643-108
89059-928PK 189.87 USD InStock
89059-928 89059-906 89084-592 89059-926 89059-924 89059-910 89059-932 89059-876 89059-930 89059-896 89059-894 10099-434 10099-436 89059-918 89084-582 89084-580 10099-430 89059-914 89059-936 10099-432 89059-912 89059-944 89059-900 89059-922 89059-920 89059-942 89084-588 89059-940 89059-880
PTFE Sleeves, 0.05 mm, Ace Glass Incorporated
NS Joint Sleeves and Connectors Joint Sleeves
PTFE sleeves are inserted between standard taper ground glass joints
Precludes contamination from lubricant or from leaks. They have a wall thickness of 0.05mm, are inert, usable to 280°C, and will withstand low to medium vacuum.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty