Would you like to log in again?
Multi-Blade, Large Scale Agitators, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
8091-06 8091-26 8091-04 8091-10 8091-02 8091-40 8091-20 8091-36 8091-34
89062-146EA 537.36 USD InStock
89062-146 89062-156 89062-148 89062-138 10099-472 10099-474 89062-152 10099-476 89062-154
Multi-Blade, Large Scale Agitators, PTFE, Ace Glass Incorporated
Stirrers Stirring Blades
PTFE agitators are designed for use with large-scale reactors with 6", 8", or 12" flanges
These heavy-duty agitators impart maximum energy and are offset at the bottom to accomodate slurries. The 19mm agitators fit drilled-type or knob-type polished glass, precision ground glass, and PTFE covered stainless steel shafts. The 28mm agitators fit heavy duty polished glass drilled shafts.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty