Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
4-Blade PTFE Agitators, Ace Glass Incorporated
Supplier: Ace Glass
8097-06 8097-26 8097-30 8097-08 8097-28 8097-04 8097-10 8097-22 8097-12 8097-24 8097-02
10099-478EA 575.63 USD InStock
10099-478 10099-488 10767-330 10099-490 10099-492 10099-480 10099-494 10099-482 10099-496 10099-484 10099-486
4-Blade PTFE Agitators, Ace Glass Incorporated
Stirrers Stirring Blades
The 4-Blade PTFE Agitator is available with either a 45° or 90° blade angle
The agitator is designed for use with glass or metal shafts, and is secured using a compression-style fitting allowing it to be positioned anywhere along the length of the shaft.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty