Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PCYOX1 Rabbit Polyclonal Antibody

Supplier: Prosci
27-767
10105-448EA 680.93 USD InStock
10105-448
Anti-PCYOX1 Rabbit Polyclonal Antibody
Antibodies
Prenylated proteins contain one of two isoprenoid lipids, either the 15-carbon farnesyl or the 20-carbon geranylgeranyl, covalently attached to cysteine residues at or near their C terminus. PCYOX1 is capable of cleaving the thioether bond of prenylcysteines. The enzyme was isolated from bovine brain membranes and exhibits an apparent molecular mass of 63 kDa. This is a specific enzyme involved in prenylcysteine metabolism in mammalian cells, most likely comprising the final step in the degradation of prenylated proteins.

PCYOX1 antibody can be used for detection of PCYOX1 by ELISA at 1:12500. PCYOX1 antibody can be used for detection of PCYOX1 by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PCYOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty