Would you like to log in again?
20X SSC, Grow Cells
Supplier: GrowCells
MRGF-6496-6 MRGF-6496 MRGF-6495-010L MRGF-6495-12
101076-122EA 284.89 USD InStock
101076-122 101076-212 101076-214 101076-120
20X SSC, Grow Cells
Buffers Electrophoresis Buffers Hybridization Buffers
Sterile 20X Saline-Sodium Citrate (SSC) prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging
Concentrated sodium chloride-sodium citrate buffer functions by providing high ionic strength to optimize the rate of probe annealing to target nucleic acid. Working solutions can easily be prepared from concentrated 20X solution with DI water.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty