Would you like to log in again?
Oakton® pH 700 Benchtop pH Meter, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Oakton®
WD-35419-12 WD-35419-10 35419-03 WD-35419-00
101076-304EA 902.44 USD InStock
101076-304 101076-306 101076-308 76221-212
Oakton® pH 700 Benchtop pH Meter, Cole-Parmer
Electrochemical Meters pH and Multi-Parameter Meters
Microprocessor based pH 700 benchtop meter measures pH, mV, relative mV, and temperature.
  • Large display permits easier readout viewing
  • The large, dual-display LCD provides excellent visibility, even from distance
  • Datalogging allows you to store and recall up to 100 data sets
  • Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures
Five point pH calibration, with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet.
Meter features HOLD, ATC and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. Meter accepts any pH or ORP electrodes with a BNC connector.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty