Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oakton® pH Buffer Solutions, Cole-Parmer

Supplier: Cole Parmer
Oakton®
05942-11 05942-44 05942-25 00653-14 WD-00654-00 WD-00654-04 05942-24 00653-10 00653-09 05942-64 00653-13 05942-42 00654-03 00654-12 05942-65 05942-41 05942-10 00653-12 05942-21 05942-45 05942-61 WD-00654-08 00654-01 00653-07 05942-22 00653-11 00654-07 00653-08 05942-62
76325-306PK 75.25 USD InStock
76325-306 76325-304 76325-402 76325-308 76325-290 101076-320 76325-294 76325-392 76325-292 76325-390 76325-298 76325-396 76325-296 76325-394 76325-312 76325-398 76325-310 76325-316 76325-314 76325-318 76325-386 101076-316 76325-320 101076-318 76325-384 76325-302 76325-400 76325-300 76325-388
Oakton® pH Buffer Solutions, Cole-Parmer
Buffers pH Calibration Buffers
Ideal for laboratory and field analysis

  • pH buffers are traceable to NIST standard reference materials
  • Buffer solution lets you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
  • pH Buffer Solutions have pH/temperature tables printed on the labels for accurate calibrations (not on 60-mL, but included as insert)
  • Manufactured under ISO 9000 quality guidelines
  • If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days

Packaging: Plastic bottles.

Caution: Store buffers away from direct sunlight.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty