Would you like to log in again?
pH Singles Calibration Pouches, Oakton Instruments
Supplier: Cole Parmer
WD-35653-04
101076-328PK 66.6 USD InStock
101076-328
pH Singles Calibration Pouches, Oakton Instruments
Buffers pH Calibration Buffers
Calibrate your pH meter in the lab or in the field with convenient, portable, and highly accurate pH Singles
  • No Unwieldy Bottles to Carry or Clean
The pouches are airtight for maximum freshness and unquestionable reliability. All solutions are certified traceable to NIST Standard Reference Materials for the highest accuracy. You get twenty 20 mL pouches, five each of pH 4, 7, and 10, and rinse water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty