Would you like to log in again?
Oakton SALT 6+ Handheld Salinity Meter, Cole-Parmer
Supplier: Cole Parmer
Oakton SALT 6+
WD-35604-44
101076-356EA 894.41 USD InStock
101076-356
Oakton SALT 6+ Handheld Salinity Meter, Cole-Parmer
Electrochemical Meters Salinity Meters
Autoranging salinity with temperature measurements.
  • Temperature readout in °C with the push of a button
  • Rubber boot with built-in stand protects meter and adds functionality
  • Hold function; auto-off function shuts meter off after 20 minutes of nonuse
These microprocessor-based meters feature easy, push-button operation that offers high accuracy at an extremely affordable price. The SALT 6+ features single-point calibration; calibrate using 4487-ppm calibration solution 00653-89 (sold separately).
Ordering information: Supplied with protective rubber boot with stand, salinity probe (K = 1.0), calibration solutions, sample bottles, rinse bottle, hard plastic carrying, and four AAA batteries.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty