Would you like to log in again?
Endo Trap® HD Leakage ELISA, Biovendor
Supplier: BioVendor
LET0014
101076-596EA 1523.37 USD InStock
101076-596
Endo Trap® HD Leakage ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of endotoxin.
EndoTrap affinity chromatography is one principal method for cleaning biological solutions from endotoxin contaminations. The EndoTrap ligand is a protein by nature, which is bound to the polymeric bead-matrix by stable covalent bonds. However, leakage of minute amounts of ligand is a matter of fact for all affinity materials and testing on these contaminants is often required for regulatory purposes. Depending on the intended use of the preparation and the step in the purification process (early or late), where EndoTrap is used, a quantitative analysis of residual EndoTrap ligand might be required. The EndoTrap Leakage ELISA was developed to allow an accurate and reproducible determination of small amounts of EndoTrap ligand in biological samples. This ELISA is suitable for detection of leached ligand from EndoTrap HD resin and EndoTrap blue resin. EndoTrap® HD is intended for in vitro quantitative removal of lipopolysaccharide (LPS) from biological samples in aquatic solutions such as proteins, antibodies, cell extracts and nucleic acids. EndoTrap® HD can be used in small scale processes for R&D and large scale processes, like manufacturing. It can be applied in early or late biomanufacturing process steps. EndoTrap® HD is based on hydrophilic, dimensionally stable affinity matrix with excellent pressure/flow characteristics. A EndoTrap® Leakage ELISA is available for the quantitative determination of the EndoTrap® ligand leakage.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty