Would you like to log in again?
Periostin, OSF2, Human ELISA, BioVendor
Supplier: BioVendor
RAG019R
101076-606EA 944.15 USD InStock
101076-606
Periostin, OSF2, Human ELISA, BioVendor
Assays ELISAs
Periostin is thought to be involved in osteoblast recruitment, attachment and spreading.
  • Calibration Range: 78–5000 pg/mL
  • Limit of Detection: 15 pg/mL
  • Cell culture supernatant, Plasma, Serum: sample diluiton will need to be determined, start at 50 to 100x
Periostin was originally isolated as an osteoblast-specific factor that functions as a cell adhesion molecule for pre-osteoblasts and is thought to be involved in osteoblast recruitment, attachment and spreading. Additionally, periostin expression has previously been shown to be significantly increased by both transforming growth factor β1 (TGF-β1) and bone morphogenetic protein 2 (BMP-2). Abnormal expression of periostin is also linked to angiogenesis and metastasis in epithelial tumors. Periostin up-regulation in cancers usually correlates with aggressiveness and/or poor survival.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty