Would you like to log in again?
Corticosterone Mouse/Rat ELISA, BioVendor
Supplier: BioVendor
RTC002R
101076-642EA 833.29 USD InStock
101076-642
Corticosterone Mouse/Rat ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of corticosterone.
  • Calibration Range: 15 - 2250 ng/ml
  • Limit of Detection: 6.1 ng/ml
  • Plasma, serum: 10 µl/well
The BioVendor Mouse/Rat Corticosterone ELISA is a competitive immunoassay for the measurement of corticosterone in rat and mouse serum or plasma. Corticosterone is secreted by the adrenal cortex under control of the pituitary hormone ACTH via a negative feedback mechanism. It is the most abundant circulating steroid in rats, since rodents are not able to synthesize Cortisol, the major glucocorticoid in human, as a result of lacking the enzyme C17-Hydroxylase.
The BioVendor Mouse/Rat Corticosterone ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. An unknown amount of corticosterone present in the sample and a defined amount of corticosterone conjugated to horseradish peroxidase compete for the binding sites of corticosterone antiserum coated to the wells of a microplate. After incubation on a shaker the microplate is washed four times. After addition of the substrate solution the concentration of corticosterone is inversely proportional to the optical density measured.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty