Would you like to log in again?
Rat/Mouse Progesterone ELISA, Biovendor
Supplier: BioVendor
RTC008R
101076-648EA 833.29 USD InStock
101076-648
Rat/Mouse Progesterone ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of progesterone.
The BioVendor Mouse/Rat Progesterone ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. An unknown amount of progesterone present in the sample and a defined amount of progesterone conjugated to horseradish peroxidase compete for the binding sites of progesterone antiserum coated to the wells of a microplate. After incubation on a shaker the microplate is washed four times. After addition of the substrate solution the concentration of progesterone is inversely proportional to the optical density measured.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty