Would you like to log in again?
Multiply® PCR Tubes, SARSTEDT
Supplier: Sarstedt
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Multiply®
72.737 72.985.002 72.737.002 72.735.002
101093-248BX 108.97 USD InStock
101093-248 101093-485 101093-188 101093-235
Multiply® PCR Tubes, SARSTEDT
Tubes PCR Tubes
These polypropylene tubes feature a flat lid, and ensure contamination-free sample collection.
  • Thin, smooth walls ensure rapid and even heat transfer to the sample
  • Anti-contamination shield prevents accidental contact with the inner surface of the lid, preventing potential contamination
  • Frosted flat lid with large writing area
Developed due to the limitations of traditional reaction tubes, Multiply® PCR Tubes offer many desirable benefits. Enhanced design features ensure work is risk-free from contamination and non-tiring compared to the more traditional tubes.
The 0.2 and 0.5 ml thin-walled Multiply® PCR cups are molded with an integral anti-contamination shield at the front of the lid.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty