Would you like to log in again?
RACK 18 PLACE GRIPPER
Supplier: SARSTEDT INC MS
18 PLACE ""GRIPPER"" RACK (3X6)^ FOR SCREW CAP MICRO TUBES 0.5-2.0ML, POLYCARBONATE, AUTOCLAVABLE.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RACK 18 PLACE GRIPPER 101093-320 93.894 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty