Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Microtube Racks, Sarstedt®

Supplier: Sarstedt
93.1428 93.837
101093-472EA 143.33 USD InStock
101093-472 101093-474
Microtube Racks, Sarstedt®
Racks Tube Racks
Racks made of transparent PC. Suitable for 48 × 1,5 ml reaction tubes or screw cap micro tubes.

  • Impact resistant material
  • Autoclavable up to 121 °C
  • Format: 12 x 4

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty