Would you like to log in again?
Micro Tubes with Thread, Type A, Sarstedt
Supplier: Sarstedt
72.730.003 72.730.007 72.703.406 72.730.008 72.730.305 72.730.306 72.730.009 72.730
101093-797PK 51.43 USD InStock
101093-797 101093-589 101093-779 101093-616 102971-720 103575-128 101093-597 101093-740
Micro Tubes with Thread, Type A, Sarstedt
Tubes Microtubes
Micro Tubes are for transport, storage and sample processing in the laboratory with safe screw cap with O-ring.
  • Autoclavable at 121 °C
  • Tubes are skirted for self-standing and neutral cap
Knurls enable single-handed operation in special Sarsetedt racks for micro tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty