Would you like to log in again?
Reagent/Centrifuge Tubes, Flat Base, with Push Cap, SARSTEDT
Supplier: Sarstedt
58.536.300
101093-720PK 69.23 USD InStock
101093-720
Reagent/Centrifuge Tubes, Flat Base, with Push Cap, SARSTEDT
Tubes Centrifuge Tubes
Centrifuge tubes are made of polypropylene with LDPE push caps.
  • Wide range of applications
Certifications: CE marked.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty