Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobenzoic acid

Supplier: USP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: PABAVitamin Bxpara-Aminobenzoic acidVitamin H1

1019803
101094-262EA 420 USD InStock
101094-262
4-Aminobenzoic acid
4-Aminobenzoic acid


Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007894
CAS Number: 150-13-0
EINECS: 205-753-0
Merck Index: 13,00422
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty