Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride dihydrate

Supplier: USP
Warning
Synonyms: Calcium Chloride Dihydrate

1086436
101094-464EA 429.23 USD InStock
101094-464
Calcium chloride dihydrate
Calcium chloride dihydrate


Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1,85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty