Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Capsaicin

Supplier: USP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: 8-Methyl-N-vanillyl-6-none-amid

1091108
101094-486EA 429.23 USD InStock
101094-486
Capsaicin
Capsaicin


Formula: C₁₈H₂₇NO₃
MW: 305.42 g/mol
Boiling Pt: 210…220 °C (0.01 mmHg)
Melting Pt: 66.1 °C
MDL Number: MFCD00017259
CAS Number: 404-86-4
EINECS: 206-969-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,01811
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty