Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cholic acid

Supplier: USP

1133503
101094-614EA 403.08 USD InStock
101094-614
Cholic acid
Cholic acid


Formula: C₂₄H₄₀O₅
MW: 408.58 g/mol
Boiling Pt: 545 °C (1013 hPa)
Melting Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00003672
CAS Number: 81-25-4
EINECS: 201-337-8
Merck Index: 13,02223
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty