Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dihydroxyacetone

Supplier: USP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Dihydroxyacetone

1204102
101094-832EA 383.08 USD InStock
101094-832
1,3-Dihydroxyacetone
1,3-Dihydroxyacetone


Formula: C₃H₆O₃
MW: 90.08 g/mol
Melting Pt: 75 °C
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004670
CAS Number: 96-26-4
EINECS: 202-494-5
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty