Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Docusate sodium

Supplier: USP
Warning
Synonyms: Sodium dioctyl sulphosuccinateSodium dioctyl sulfosuccinateDioctyl sulfosuccinate sodium saltbis(2-Ethylhexyl) sodium sulfosuccinateDioctyl sulphosuccinate, sodium saltDioctyl sulphosuccinate sodium saltbis(2-Ethylhexyl) sodium sulphosuccinate

1224802
101094-868EA 513.85 USD InStock
101094-868
Docusate sodium
Docusate sodium


Formula: C₂₀H₃₇NaO₇S
MW: 444.57 g/mol
Boiling Pt: ∼180 °C (1013 hPa)
Melting Pt: ∼155 °C
Density: ∼1,1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012455
CAS Number: 577-11-7
EINECS: 209-406-4
Merck Index: 12,03460
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty