Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guaiacol

Supplier: USP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: 2-Methoxyphenol2-Hydroxyanisole1-Hydroxy-2-methoxybenzene

1300004
101095-102EA 407.69 USD InStock
101095-102
Guaiacol
Guaiacol


Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 28…32 °C
Density: 1.1534 g/cm³ (0 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002185
CAS Number: 90-05-1
EINECS: 201-964-7
UN: 2810
ADR: 6.1,III
Merck Index: 13,04568
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty