Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrocortisone

Supplier: USP

1316004
101095-150EA 420 USD InStock
101095-150
Hydrocortisone
Hydrocortisone


Formula: C₂₁H₃₀O₅
MW: 362.47 g/mol
Boiling Pt: 567 °C (1013 hPa)
Melting Pt: 202…204 °C
Density: 1,4 g/cm³ (20 °C)
MDL Number: MFCD00011654
CAS Number: 50-23-7
EINECS: 200-020-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty