Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitroglycerin 1% in propylene glycol USP Reference Standard

Supplier: USP
Danger

1466506
101095-632EA 427.69 USD InStock
101095-632
Nitroglycerin 1% in propylene glycol USP Reference Standard
Nitroglycerin
Diluted nitroglycerin each ampules, approximately have 200 mg of a 0.974% solution in propylene glycol.


Formula: C₃H5N₃O₉
CAS Number: 55-63-0
UN: 1204
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty