Would you like to log in again?
ImmPrint™ Permanent Marking Pen, Vector Laboratories
Supplier: Vector Laboratories
H-6100
101098-070EA 138.46 USD InStock
101098-070
ImmPrint™ Permanent Marking Pen, Vector Laboratories
Pens Lab Markers
An all-purpose marking pen that's designed for writing on slides, with marks that remain legible through histology processing.
  • Solvent-resistant permanent marking pen
  • Designed for writing on frosted glass microscope slides, tissue cassettes, and most hard surfaces
  • High density, fast-drying, black ink is resistant to most organic solvents
  • Has a smooth writing tip that resists drying out
The ImmPrint™ Histology Pen is a solvent-resistant permanent marking pen designed for writing on frosted glass microscope slides, tissue cassettes, and most hard surfaces. The high density, fast-drying, black ink is resistant to most organic solvents encountered in histological applications such as alcohols and clearing agents. Unlike other pens commonly used for histology, the ImmPrint™ pen has a smooth writing tip that resists drying out.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty