Would you like to log in again?
VECTASTAIN® ABC-Peroxidase Kit, Vector Laboratories
Supplier: Vector Laboratories
PK-4010 PK-4000 PK-4007 PK-4004 PK-4002 PK-4005 PK-4001
101098-304EA 720 USD InStock
101098-304 101098-312 101098-302 101098-298 101098-296 101098-294 101098-308
VECTASTAIN® ABC-Peroxidase Kit, Vector Laboratories
Blotting Kits
VECTASTAIN® ABC kits take advantage of the flexibility and stability of avidin/biotin binding for sensitive, cost-effective chromogenic target protein detection on tissue slices and blots.
  • Avidin/biotin complex (ABC) kits offer a high level of sensitivity and flexibility
  • The VECTASTAIN® ABC-HRP Standard Kit may be used to detect any biotinylated target, not just secondary antibodies
  • Relatively low-cost protein detection
With more than 50000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories. The ABC kit system is modular and different variations can accommodate a wide array of primary antibody and tissue species. Kits use horseradish peroxidase (HRP) as a detection enzyme. Several kits also use a secondary antibody to target primary antibodies. The ABC complex, once formed, is stable and can be used for several days after preparation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty