Would you like to log in again?
VECTASTAIN® Elite® ABC-Peroxidase Kit, Vector Laboratories
Supplier: Vector Laboratories
PK-6105 PK-7100 PK-6200 PK-6103 PK-6106 PK-6101 PK-6102 PK-6104 PK-6100 PK-7200
101098-314EA 456.92 USD InStock
101098-314 101098-326 101098-324 101098-332 101098-322 101098-330 101098-320 101098-318 101098-316 101098-328
VECTASTAIN® Elite® ABC-Peroxidase Kit, Vector Laboratories
Blotting Kits
This VECTASTAIN® Elite® ABC system is the most sensitive avidin/biotin-based peroxidase system and is particularly important in the localization of antigens present in low amounts or in cases where the cost of the primary antibodies is significant.
  • Smaller, very uniform, and highly active, allowing more accessibility for binding to a biotinylated target
  • Higher sensitivity leads to lower cost per slide
  • Used to detect any molecule that is biotinylated
With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories. The ABC kit system is modular and different variations can accommodate a wide array of primary antibody and tissue species. This kit uses horseradish peroxidase (HRP) in a compact but highly active ABC detection complex. Several kits also use a secondary antibody to target primary antibodies. The ABC complex, once formed, is stable and can be used for several days after preparation.
Delivery information: Kit includes Reagent A (Avidin DH) 2 mL and reagent B (biotinylated horseradish peroxidase H) 2 mL sufficient to stain 500 to 1000 tissue sections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty