Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Avidin/Biotin Blocking Kit, Vector Laboratories
Supplier: Vector Laboratories
SP-2001
101098-512EA 206.31 USD InStock
101098-512
Avidin/Biotin Blocking Kit, Vector Laboratories
Electrophoresis Reagents Western Blotting Reagents
Avidin/Biotin Blocking Kit blocks all endogenous biotin, biotin receptors, and avidin binding sites present in tissues.
Certifications: CE marked.
Delivery information: Kit includes Avidin D (18 mL) and biotin (18 mL) in dropper bottles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty