Would you like to log in again?
Cryogenic Vials, Internal or External Threaded, Phenix Research Products
Supplier: Phenix
C-411F C-402F C-403F C-414F C-401F C-405F
101100-384PK 60.82 USD InStock
101100-384 101100-374 101100-210 101100-386 101100-134 101100-084
Cryogenic Vials, Internal or External Threaded, Phenix Research Products
Vials
These freestanding heavy-duty sterile cryovials have thick silicone O-ring sets.
  • Fast thread design opens or closes in 1¹/₄ turns
  • Ridged bottom for one-handed open/close
  • Ideal for use in gas phase of LN₂ only
Temperature range : −196 to +121 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty