Would you like to log in again?
Dry Block Incubator, SPS Medical
Supplier: Crosstex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NDB-036
101101-846EA 265.38 USD InStock
101101-846
Dry Block Incubator, SPS Medical
Block Heaters Digital Block Heaters
Specifically designed for the STERRAD (hydrogen peroxide, gas plasma sterilization process)
Pre-set at 56ºC for incubating Geobacillus stearothermophilus or 37ºC for Bacillus atrophaeus. Each incubator is calibrated to +2ºC and comes with a lifetime warranty.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty