Would you like to log in again?
m-Endo Media Ampoules, Advantec
Supplier: Advantec
COLI200
101105-196PK 136.32 USD InStock
101105-196
m-Endo Media Ampoules, Advantec
Cell Culture Supplements and Additives
m-Endo Media Ampules are used with membrane filters to culture bacteria, yeasts, and molds
  • Single use vials assure sterility
  • Culture bacteria, yeasts and molds
  • Certified to comply with established test criteria
Available ready-to-use 2 mL ampoules contain enough medium to saturate one standard absorbent pad in a petri dish. Single use vials assure sterility. Each product is lot tested and certified to comply with established test criteria.
Delivery information: Quality certificates are included with each shipment, and packages are clearly marked with expiration date.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty