Would you like to log in again?
Pressure Vessels, Stainless Steel, Advantec MFS
Supplier: Advantec
720160 720360 740160 741640 731040 740560 751660 741060 730840 740860 731060 741040 720540 740140 720560 731660 720340 730860 720140 740340 740840 740360 740540
101105-828PK 5380.28 USD InStock
101105-828 101105-838 101105-826 101105-804 101105-834 101105-890 101105-800 101105-822 101105-888 101105-832 101105-798 101105-764 101105-830 101105-840 101105-246 101105-796 101105-884 101105-872 101105-850 101105-750 101105-882 101105-650 101105-892
Pressure Vessels, Stainless Steel, Advantec MFS
Reaction Vessels Reaction Vessels, Cylindrical Shape
Advantec Pressure Vessels are used to contain process fluids for filtration through pressure type filter holders
Use under positive pressure or negative pressure. When working with negative pressure or operating at low pressure(<30psi), use the replacement vacuum closure.
Type 316/316L stainless steel complies with good manufacturing practices and is recommended for low pH and high salt applications. Type 304 stainless steel Standard Closure vessels offer 155psi (1–5 gallons) or 115psi (8–16 gallons) maximum operating pressure at 149°C (300°F). Type 304 stainless steel Wide Mouth Closure vessels offer 125psi (1–5 gallons) or 110psi (8–16 gallons) maximum operating pressure at 149°C (300°F). Type 316L stainless steel Standard Closure vessels offer 150psi (1–5 gallons) or 110psi (8–16 gallons) maximum operating pressure at 149°C (300°F). Type 316L stainless steel Wide Mouth Closure vessels offer 125psi (1–5 gallons) or 110psi (8–16 gallons) maximum operating pressure at 149°C (300°F).
The pressure vessels are available in type 304 or 316L stainless steel. Choose either standard closure or wide mouth closure (for ease of cleaning). Electropolished finish provides a smooth surface for cleaning. Molded rubber skirt/base protects vessel against rough handling (also available with stainless base for continuous operation at high temperatures).
Certifications: ASME compliant.
Ordering information: Accessories not included. Optional O-ring materials and replacement parts available.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty