Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P4 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-010-100 905-010
101106-226EA 342 USD InStock
CA101106-226 101106-226
Anti-P4 Mouse Monoclonal Antibody
Antibodies
Progesterone is the major female sex hormone, and is responsible for reproductive-related activities such as breast glandular development, the endometrial aspects of the menstrual cycle, and the establishment and maintenance of pregnancy. In addition, progesterone also directs pregnancy support physiology including changes in carbohydrate, protein and lipid metabolism, thermoregulation, sodium reabsorption in renal tubules and the reduction of alveolar and atrial carbon dioxide partial pressures (PCO2). Progesterone is secreted in large amounts by the corpus luteum and by the extracellular conversion of cholesterol, cholestryl esters, adrenal steroids, prenenolone and pregnenolone sulfate. Small quantities are also secreted directly from the adrenal glands.

  • For ELISA, Flow Cytometry

Species independent

Type: Primary
Antigen: P4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty