Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-12(L)-HETE Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-053-100 905-053
101106-274EA 385 USD InStock
CA101106-274 101106-274
Anti-12(L)-HETE Rabbit Polyclonal Antibody
Antibodies
HETEs are byproducts generated by the metabolism of arachidonic acid by lipoxygenases. 12(S)-HETE is the stereospecific hydroxy product from the reduction of 12(S)-hydroperoxy tetraenoic eicosatetraenoic acid [12(S)-HpETE], which itself is a 12-lipoxygenase metabolite of arachidonic acid. 12(S)-HETE is chemotactic and chemokinetic for polymorphonuclear leukocytes and vascular smooth muscle cells. It also acts as a second messenger in angiotensin-II induced aldosterone production. 15-hydroxyeicosatetraenoic acid [15(S)-HETE] is the major hydroxy derivative of arachidonic acid when acted upon by 15-lipoxygenase. It is also the primary monohydroxy acid synthesized by the lipoxygenase activity of Cyclooxygenase-1 upon linoleic acid. 15(S)-HETE acts as a paracrine regulator of smooth muscle and lung neutrophil recruitment, and elevated levels of 15(S)-HETE are associated with asthma, rhinitis, chronic paranasal sinusitis and rheumatoid arthritis.

  • For ELISA, Flow Cytometry

Species independent.

Type: Primary
Antigen: 12(L)-HETE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty